Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGCCGTGTATCCCCTCCTTGTC[C/G]CAGCTCCTGCCCCTGCCGGGGCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 124015 MIM: 616695 MIM: 616550 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
POR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POR - cytochrome p450 oxidoreductase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STYXL1 - serine/threonine/tyrosine interacting-like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM120A - transmembrane protein 120A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317803.1 | Intron | NP_001304732.1 | ||||
NM_031925.2 | Intron | NP_114131.1 |