Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGGTACCTACTCAGAACCTAAAT[C/T]GGTAAGCTTATTTTTTCTCTCTTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604741 | ||||||||||||||||||||
Literature Links: |
AKR1D1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AKR1D1 - aldo-keto reductase family 1, member D1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190906.1 | 179 | Missense Mutation | TCG,TTG | S,L 31 | NP_001177835.1 | |
NM_001190907.1 | 179 | Missense Mutation | TCG,TTG | S,L 31 | NP_001177836.1 | |
NM_005989.3 | 179 | Missense Mutation | TCG,TTG | S,L 31 | NP_005980.1 |