Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCTGCCTCACCAAGGCGAAGAG[G/T]GAGGAGTAGAAACGCTGATGTTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615054 MIM: 603354 | ||||||||||||||||||||
Literature Links: |
ASB10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASB10 - ankyrin repeat and SOCS box containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142459.1 | 1389 | Silent Mutation | TCA,TCC | S,S 414 | NP_001135931.2 | |
NM_001142460.1 | 1389 | Silent Mutation | TCA,TCC | S,S 376 | NP_001135932.2 | |
NM_080871.3 | 1389 | Silent Mutation | TCA,TCC | S,S 399 | NP_543147.2 |
GBX1 - gastrulation brain homeobox 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |