Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACAGATCTCCAACTGGTTACCCC[A/G]GCGCTTTAAAACCAGCTGGTCAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611986 MIM: 610337 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPS24 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPS24 - mitochondrial ribosomal protein S24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032014.2 | 388 | Missense Mutation | NP_114403.1 |
URGCP - upregulator of cell proliferation | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
URGCP-MRPS24 - URGCP-MRPS24 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204871.1 | 388 | UTR 3 | NP_001191800.1 |