Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGCTGATGGCCGCTGACAGGGG[A/G]CGCAGGTAAGTAGGGAACGGTCTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600639 MIM: 616524 | ||||||||||||||||||||
Literature Links: |
CASP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CASP2 - caspase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001224.4 | 310 | Intron | NP_001215.1 | |||
NM_032982.3 | 310 | Silent Mutation | GGA,GGG | G,G 23 | NP_116764.2 | |
NM_032983.3 | 310 | Silent Mutation | GGA,GGG | G,G 23 | NP_116765.2 |
LOC105375545 - uncharacterized LOC105375545 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM139 - transmembrane protein 139 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242773.2 | 310 | Intron | NP_001229702.1 | |||
NM_001242774.2 | 310 | Intron | NP_001229703.1 | |||
NM_001242775.2 | 310 | Intron | NP_001229704.1 | |||
NM_001282876.1 | 310 | Intron | NP_001269805.1 | |||
NM_001282877.1 | 310 | Intron | NP_001269806.1 | |||
NM_153345.3 | 310 | Intron | NP_699176.1 |