Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604019 MIM: 609097 | ||||||||||||||||||||
Literature Links: |
AGFG2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AGFG2 - ArfGAP with FG repeats 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FBXO24 - F-box protein 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRCH4 - leucine rich repeats and calponin homology domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289934.1 | 2051 | Intron | NP_001276863.1 | |||
NM_002319.4 | 2051 | Missense Mutation | CCC,TCC | P,S 655 | NP_002310.2 |
SAP25 - Sin3A associated protein 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZASP - ZO-2 associated speckle protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289933.1 | 2051 | Intron | NP_001276862.1 |