Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCACTTCCTCCTGCTCTATCAGAA[A/G]AAGGGGCAGTGGTACGAGATCTACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601232 | ||||||||||||||||||||
Literature Links: |
PIK3CG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PIK3CG - phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282426.1 | 449 | Silent Mutation | AAA,AAG | K,K 102 | NP_001269355.1 | |
NM_001282427.1 | 449 | Silent Mutation | AAA,AAG | K,K 102 | NP_001269356.1 | |
NM_002649.3 | 449 | Silent Mutation | AAA,AAG | K,K 102 | NP_002640.2 | |
XM_005250443.3 | 449 | Silent Mutation | AAA,AAG | K,K 102 | XP_005250500.1 | |
XM_011516316.1 | 449 | Silent Mutation | AAA,AAG | K,K 102 | XP_011514618.1 | |
XM_011516317.1 | 449 | Silent Mutation | AAA,AAG | K,K 102 | XP_011514619.1 | |
XM_017012328.1 | 449 | Silent Mutation | AAA,AAG | K,K 102 | XP_016867817.1 |