Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAATAAGAAAACATTCACTACCT[C/T]TTGCACATGCACGTTTGCAATCCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600033 | ||||||||||||||||||||
Literature Links: |
LOC105375401 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105375401 - uncharacterized LOC105375401 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TFPI2 - tissue factor pathway inhibitor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271003.1 | 866 | Missense Mutation | AAA,AGA | K,R 199 | NP_001257932.1 | |
NM_001271004.1 | 866 | Missense Mutation | AAG,GAG | K,E 185 | NP_001257933.1 | |
NM_006528.3 | 866 | Missense Mutation | AAA,AGA | K,R 210 | NP_006519.1 |