Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGCCCCTGAGCAGCTGCTGAGTC[A/T]GGCCCGGGACTTCATCAACCAGTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 163729 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOS3 - nitric oxide synthase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000603.4 | 430 | Missense Mutation | CAG,CTG | Q,L 126 | NP_000594.2 | |
NM_001160109.1 | 430 | Missense Mutation | CAG,CTG | Q,L 126 | NP_001153581.1 | |
NM_001160110.1 | 430 | Missense Mutation | CAG,CTG | Q,L 126 | NP_001153582.1 | |
NM_001160111.1 | 430 | Missense Mutation | CAG,CTG | Q,L 126 | NP_001153583.1 | |
XM_006716002.3 | 430 | Missense Mutation | CAG,CTG | Q,L 126 | XP_006716065.1 | |
XM_017012232.1 | 430 | Missense Mutation | CAG,CTG | Q,L 126 | XP_016867721.1 | |
XM_017012233.1 | 430 | UTR 5 | XP_016867722.1 | |||
XM_017012234.1 | 430 | Missense Mutation | CAG,CTG | Q,L 126 | XP_016867723.1 |