Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTTCCAGCCTGGCTCTGCTGGGG[C/T]GCTGGCATCTGGTTCAGTTCCACCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZC3HC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZC3HC1 - zinc finger C3HC-type containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282190.1 | 1941 | Missense Mutation | NP_001269119.1 | |||
NM_001282191.1 | 1941 | Missense Mutation | NP_001269120.1 | |||
NM_016478.4 | 1941 | Missense Mutation | NP_057562.3 | |||
XM_005250403.2 | 1941 | Missense Mutation | XP_005250460.1 | |||
XM_011516288.2 | 1941 | Missense Mutation | XP_011514590.1 | |||
XM_011516289.2 | 1941 | Missense Mutation | XP_011514591.1 | |||
XM_011516290.2 | 1941 | Missense Mutation | XP_011514592.1 | |||
XM_017012287.1 | 1941 | Missense Mutation | XP_016867776.1 | |||
XM_017012288.1 | 1941 | Missense Mutation | XP_016867777.1 |