Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATAGTGATTGACTTCCTCCTCGGT[A/G]GGGTGGAGGCGCTGGGGGACGGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610184 MIM: 603066 | ||||||||||||||||||||
Literature Links: |
MOGAT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MOGAT3 - monoacylglycerol O-acyltransferase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287147.1 | 1078 | Missense Mutation | CAC,TAC | H,Y 236 | NP_001274076.1 | |
NM_178176.3 | 1078 | Silent Mutation | CCC,CCT | P,P 303 | NP_835470.1 | |
XM_005250309.3 | 1078 | Missense Mutation | CCA,CTA | P,L 332 | XP_005250366.1 | |
XM_011516126.2 | 1078 | Silent Mutation | CCC,CCT | P,P 267 | XP_011514428.1 | |
XM_011516127.2 | 1078 | Silent Mutation | CCC,CCT | P,P 185 | XP_011514429.1 |
PLOD3 - procollagen-lysine,2-oxoglutarate 5-dioxygenase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |