Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCAGGACATAGGGATGCCCAAG[A/G]AGACAGGCTCCAGTTCTACCGTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 118425 | ||||||||||||||||||||
Literature Links: |
CLCN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLCN1 - chloride voltage-gated channel 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000083.2 | 186 | Missense Mutation | AAG,GAG | K,E 81 | NP_000074.2 | |
XM_011515782.2 | 186 | Intron | XP_011514084.1 | |||
XM_017011739.1 | 186 | UTR 5 | XP_016867228.1 | |||
XM_017011740.1 | 186 | UTR 5 | XP_016867229.1 |