Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGTCTTGCTAATTCTTGAGCTTT[A/G]TCTTCTAAGTATCTTTGATTGTCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 238331 MIM: 150240 | ||||||||||||||||||||
Literature Links: |
DLD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DLD - dihydrolipoamide dehydrogenase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LAMB1 - laminin subunit beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002291.2 | 5603 | Silent Mutation | GAC,GAT | D,D 1756 | NP_002282.2 | |
XM_017012201.1 | 5603 | Silent Mutation | GAC,GAT | D,D 1780 | XP_016867690.1 | |
XM_017012202.1 | 5603 | Intron | XP_016867691.1 |