Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGACCCCCCTCCCTTTTTGGAAT[A/G]ATGGGGATCTTTTTGGTGTATGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
VSTM2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VSTM2A - V-set and transmembrane domain containing 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301009.1 | 409 | Missense Mutation | ATA,ATG | I,M 1 | NP_001287938.1 | |
NM_001317843.1 | 409 | Missense Mutation | ATA,ATG | I,M 1 | NP_001304772.1 | |
NM_182546.3 | 409 | Missense Mutation | ATA,ATG | I,M 1 | NP_872352.2 | |
XM_006715665.3 | 409 | Missense Mutation | ATA,ATG | I,M 1 | XP_006715728.1 | |
XM_006715666.3 | 409 | Missense Mutation | ATA,ATG | I,M 1 | XP_006715729.1 | |
XM_017011841.1 | 409 | Missense Mutation | ATA,ATG | I,M 1 | XP_016867330.1 |