Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGCCAAAAGCTGAGTAGTGAGG[C/T]TGCCGCTGGTCCAGGAGAGGGTGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600011 MIM: 602372 | ||||||||||||||||||||
Literature Links: |
EPHB4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EPHB4 - EPH receptor B4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004444.4 | 2857 | Silent Mutation | CAA,CAG | Q,Q 903 | NP_004435.3 | |
XM_017011816.1 | 2857 | Silent Mutation | CAA,CAG | Q,Q 921 | XP_016867305.1 |
ZAN - zonadhesin (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |