Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAGTGGAAGGACAGTGAGGACCC[A/G]GACGTGCCTGCTATATGGCTCGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147290 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INHBA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INHBA - inhibin beta A subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017012174.1 | 3093 | Silent Mutation | TCC,TCT | S,S 484 | XP_016867663.1 | |
XM_017012175.1 | 3093 | Silent Mutation | TCC,TCT | S,S 419 | XP_016867664.1 | |
XM_017012176.1 | 3093 | Silent Mutation | TCC,TCT | S,S 362 | XP_016867665.1 |
INHBA-AS1 - INHBA antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |