Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGGTGGCGACGGGCATCACGGAC[A/G]CTGGAGCCTGGTAGGGCCGCCGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615778 MIM: 609003 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLDN15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLDN15 - claudin 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185080.1 | 973 | Missense Mutation | GCG,GTG | A,V 203 | NP_001172009.1 | |
NM_014343.2 | 973 | Missense Mutation | GCG,GTG | A,V 203 | NP_055158.1 |
FIS1 - fission, mitochondrial 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNHIT1 - zinc finger HIT-type containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |