Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCTCCACGTCAATGTCATAGCA[C/T]GCCGTCTTCTTCTGGTCTGAAGGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608037 MIM: 615245 MIM: 601737 | ||||||||||||||||||||
Literature Links: |
CHPF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHPF2 - chondroitin polymerizing factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR671 - microRNA 671 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMARCD3 - SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003801.1 | 1158 | Silent Mutation | GCA,GCG | A,A 356 | NP_001003801.1 | |
NM_001003802.1 | 1158 | Silent Mutation | GCA,GCG | A,A 343 | NP_001003802.1 | |
NM_003078.3 | 1158 | Silent Mutation | GCA,GCG | A,A 343 | NP_003069.2 | |
XM_011516521.2 | 1158 | Silent Mutation | GCA,GCG | A,A 254 | XP_011514823.1 | |
XM_017012556.1 | 1158 | Silent Mutation | GCA,GCG | A,A 191 | XP_016868045.1 |