Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGCTTTTTGGTGACCAGATCTT[A/G]GACAAGCGGAACTTGGACTTCTTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MICALL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MICALL2 - MICAL like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182924.3 | 2797 | Silent Mutation | TCC,TCT | S,S 891 | NP_891554.1 | |
XM_011515520.2 | 2797 | Nonsense Mutation | CAA,TAA | Q,* 890 | XP_011513822.1 | |
XM_011515522.2 | 2797 | Nonsense Mutation | CAA,TAA | Q,* 876 | XP_011513824.1 | |
XM_011515523.2 | 2797 | Nonsense Mutation | CAA,TAA | Q,* 807 | XP_011513825.1 | |
XM_011515524.2 | 2797 | Nonsense Mutation | CAA,TAA | Q,* 319 | XP_011513826.1 | |
XM_017012619.1 | 2797 | Intron | XP_016868108.1 |