Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCTGTACAGCCTCAGCGTCCTCT[A/G]CAAAGGCGAGGCCAAGGTGGTGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606209 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
YKT6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
YKT6 - YKT6 v-SNARE homolog (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006555.3 | 187 | Missense Mutation | TAC,TGC | Y,C 10 | NP_006546.1 | |
XM_005249582.4 | 187 | Missense Mutation | TAC,TGC | Y,C 10 | XP_005249639.1 | |
XM_017011682.1 | 187 | Missense Mutation | TAC,TGC | Y,C 10 | XP_016867171.1 |