Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTTGTCTTTGCAGATGACAGTAG[C/T]AACGGCCACCTCCTACCGGAGATCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PRSS37 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRSS37 - protease, serine 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008270.2 | 2164 | Missense Mutation | ACT,GCT | T,A 194 | NP_001008271.2 | |
NM_001171951.1 | 2164 | Missense Mutation | ACT,GCT | T,A 193 | NP_001165422.1 | |
XM_005249946.4 | 2164 | Missense Mutation | ACT,GCT | T,A 224 | XP_005250003.1 | |
XM_005249947.3 | 2164 | Missense Mutation | ACT,GCT | T,A 224 | XP_005250004.1 | |
XM_011515792.2 | 2164 | Missense Mutation | ACT,GCT | T,A 161 | XP_011514094.1 | |
XM_011515793.2 | 2164 | Missense Mutation | ACT,GCT | T,A 161 | XP_011514095.1 |