Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCAATGGGACAGGTGAGCTCTC[A/G]GAGCACCAGCAGTACGTGATCGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601805 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C7orf50 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C7orf50 - chromosome 7 open reading frame 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134395.1 | 910 | Intron | NP_001127867.1 | |||
NM_001134396.1 | 910 | Intron | NP_001127868.1 | |||
NM_001318252.1 | 910 | Intron | NP_001305181.1 | |||
NM_032350.5 | 910 | Intron | NP_115726.1 | |||
XM_006715793.2 | 910 | Intron | XP_006715856.1 | |||
XM_011515580.2 | 910 | Intron | XP_011513882.1 | |||
XM_011515581.2 | 910 | Intron | XP_011513883.1 | |||
XM_011515582.2 | 910 | Intron | XP_011513884.1 | |||
XM_011515583.2 | 910 | Intron | XP_011513885.1 | |||
XM_011515584.2 | 910 | Intron | XP_011513886.1 | |||
XM_017012719.1 | 910 | Intron | XP_016868208.1 | |||
XM_017012720.1 | 910 | Intron | XP_016868209.1 | |||
XM_017012721.1 | 910 | Intron | XP_016868210.1 | |||
XM_017012722.1 | 910 | Intron | XP_016868211.1 |
GPER1 - G protein-coupled estrogen receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039966.1 | 910 | Silent Mutation | TCA,TCG | S,S 50 | NP_001035055.1 | |
NM_001098201.1 | 910 | Silent Mutation | TCA,TCG | S,S 50 | NP_001091671.1 | |
NM_001505.2 | 910 | Silent Mutation | TCA,TCG | S,S 50 | NP_001496.1 |