Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTCACACGGTGTAAGTTTTGAC[A/G]GACTTATAGAGGGGCGGGAGCAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM130 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM130 - transmembrane protein 130 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134450.1 | 1134 | Silent Mutation | TCC,TCT | S,S 429 | NP_001127922.1 | |
NM_001134451.1 | 1134 | Silent Mutation | TCC,TCT | S,S 315 | NP_001127923.1 | |
NM_152913.2 | 1134 | Silent Mutation | TCC,TCT | S,S 417 | NP_690877.1 |