Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAACGTAGAATGTCAAGCCAATC[A/G]TTAAGTCATAATGGTAGACAGTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600917 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP1R3A - protein phosphatase 1 regulatory subunit 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002711.3 | 2861 | Missense Mutation | ACG,ATG | T,M 1095 | NP_002702.2 | |
XM_005250473.3 | 2861 | Missense Mutation | ACG,ATG | T,M 894 | XP_005250530.1 |