Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGGCGACTGCCATGAACTCCGGA[G/T]CCATGAGGATCCACAGTAAAGGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604281 MIM: 610821 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DBF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DBF4 - DBF4 zinc finger | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318060.1 | 517 | UTR 5 | NP_001304989.1 | |||
NM_001318061.1 | 517 | UTR 5 | NP_001304990.1 | |||
NM_001318062.1 | 517 | UTR 5 | NP_001304991.1 | |||
NM_006716.3 | 517 | Missense Mutation | GCC,TCC | A,S 5 | NP_006707.1 |
SLC25A40 - solute carrier family 25 member 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018843.3 | 517 | Intron | NP_061331.2 |