Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTCTCACATACTGGGGCAGTCG[A/G]TGGGCTGAGAGAGCTCTTTGGATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADCK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADCK2 - aarF domain containing kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052853.3 | 1146 | Missense Mutation | GAT,GGT | D,G 248 | NP_443085.2 | |
XM_006716170.3 | 1146 | Missense Mutation | GAT,GGT | D,G 248 | XP_006716233.1 | |
XM_011516675.2 | 1146 | Missense Mutation | GAT,GGT | D,G 248 | XP_011514977.1 |