Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGATCTCACTCTGGACTGTGGT[C/G]GCTGCCATTCAGGCTGTGGAGAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603397 | ||||||||||||||||||||
Literature Links: |
ZNF282 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF282 - zinc finger protein 282 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303481.1 | 434 | Silent Mutation | GTC,GTG | V,V 102 | NP_001290410.1 | |
NM_003575.3 | 434 | Silent Mutation | GTC,GTG | V,V 102 | NP_003566.1 | |
XM_006716151.3 | 434 | Silent Mutation | GTC,GTG | V,V 102 | XP_006716214.1 |