Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCTCCTCCCCGCCCTCCTCTGT[C/G]CCCTCGGCCTCCTTCCGCCGGTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603531 MIM: 615783 MIM: 602186 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP1S1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP1S1 - adaptor related protein complex 1 sigma 1 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4653 - microRNA 4653 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAT16 - N-acetyltransferase 16 (putative) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VGF - VGF nerve growth factor inducible | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003378.3 | 1520 | Silent Mutation | GGC,GGG | G,G 434 | NP_003369.2 | |
XM_005250561.4 | 1520 | Silent Mutation | GGC,GGG | G,G 434 | XP_005250618.1 | |
XM_011516549.2 | 1520 | Silent Mutation | GGC,GGG | G,G 434 | XP_011514851.1 | |
XM_017012579.1 | 1520 | Silent Mutation | GGC,GGG | G,G 434 | XP_016868068.1 |