Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGTCATACCCACGATCATATCCT[C/G]TATCATAGTAAGAATCTCGACGTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602718 | ||||||||||||||||||||
Literature Links: |
TRA2A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRA2A - transformer 2 alpha homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282757.1 | Intron | NP_001269686.1 | ||||
NM_001282758.1 | Intron | NP_001269687.1 | ||||
NM_001282759.1 | Intron | NP_001269688.1 | ||||
NM_013293.4 | Intron | NP_037425.1 | ||||
XM_005249725.1 | Intron | XP_005249782.1 | ||||
XM_011515331.1 | Intron | XP_011513633.1 | ||||
XM_017012089.1 | Intron | XP_016867578.1 | ||||
XM_017012090.1 | Intron | XP_016867579.1 |