Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCTCTATAATATATGTCATAGA[A/T]TCTCTCTTGGGCCTGGCGTGGGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118425 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLCN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLCN1 - chloride voltage-gated channel 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM131B - family with sequence similarity 131 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031690.2 | 4165 | UTR 3 | NP_001026860.2 | |||
NM_001278297.1 | 4165 | UTR 3 | NP_001265226.1 | |||
NM_014690.4 | 4165 | UTR 3 | NP_055505.3 | |||
XM_005250073.3 | 4165 | UTR 3 | XP_005250130.1 | |||
XM_006716186.2 | 4165 | UTR 3 | XP_006716249.1 |