Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCTCTGTGGACTGTCCCACTCTG[A/G]AGTTTGAGTATGGAGATGCAGATGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616520 | ||||||||||||||||||||
Literature Links: |
AHCYL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AHCYL2 - adenosylhomocysteinase like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STRIP2 - striatin interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134336.1 | 194 | Missense Mutation | AAG,GAG | K,E 52 | NP_001127808.1 | |
NM_020704.2 | 194 | Missense Mutation | AAG,GAG | K,E 52 | NP_065755.1 | |
XM_006716069.3 | 194 | Missense Mutation | AAG,GAG | K,E 52 | XP_006716132.1 | |
XM_011516432.2 | 194 | Intron | XP_011514734.1 | |||
XM_011516435.1 | 194 | Missense Mutation | AAG,GAG | K,E 52 | XP_011514737.1 | |
XM_017012470.1 | 194 | Missense Mutation | AAG,GAG | K,E 52 | XP_016867959.1 | |
XM_017012471.1 | 194 | Intron | XP_016867960.1 | |||
XM_017012472.1 | 194 | UTR 5 | XP_016867961.1 |