Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCAGCGCGCACAGCGCCAGCCCC[A/G]TCCAGGATGCGGTCCTGAAGCGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615137 | ||||||||||||||||||||
Literature Links: |
WBSCR17 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WBSCR17 - Williams-Beuren syndrome chromosome region 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022479.2 | 564 | Missense Mutation | ATC,GTC | I,V 59 | NP_071924.1 | |
XM_011516467.2 | 564 | Missense Mutation | ATC,GTC | I,V 59 | XP_011514769.1 | |
XM_011516469.2 | 564 | Missense Mutation | ATC,GTC | I,V 59 | XP_011514771.1 | |
XM_017012521.1 | 564 | Missense Mutation | ATC,GTC | I,V 59 | XP_016868010.1 |