Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTCTTGACAGGCTTGGTTTCAAC[A/G]GTTCTACTGAGGGACATGACAGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613138 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSPAN12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TSPAN12 - tetraspanin 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012338.3 | 714 | Missense Mutation | CCG,CTG | P,L 278 | NP_036470.1 | |
XM_005250239.2 | 714 | Missense Mutation | CCG,CTG | P,L 278 | XP_005250296.1 | |
XM_011515993.1 | 714 | Missense Mutation | CCG,CTG | P,L 278 | XP_011514295.1 | |
XM_011515994.1 | 714 | Missense Mutation | CCG,CTG | P,L 278 | XP_011514296.1 | |
XM_017011912.1 | 714 | Missense Mutation | CCG,CTG | P,L 278 | XP_016867401.1 | |
XM_017011913.1 | 714 | Missense Mutation | CCG,CTG | P,L 253 | XP_016867402.1 | |
XM_017011914.1 | 714 | Missense Mutation | CCG,CTG | P,L 151 | XP_016867403.1 |