Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCAGCATGCAGTGAGCACTCAG[C/T]GATGGCCAAGTATGGGAGAAGCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM83A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM83A - family with sequence similarity 83 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288587.2 | 32 | Intron | NP_001275516.1 | |||
NM_001321630.1 | 32 | Intron | NP_001308559.1 | |||
NM_032899.6 | 32 | UTR 5 | NP_116288.2 | |||
NM_207006.2 | 32 | UTR 5 | NP_996889.1 | |||
XM_005251087.2 | 32 | Intron | XP_005251144.1 | |||
XM_011517338.1 | 32 | Intron | XP_011515640.1 |