Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGTGGGCCACTCTCCACATAGAG[C/T]GCATTCTTTGCCACTGGTGGCTAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 170993 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105375907 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105375907 - uncharacterized LOC105375907 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PEX2 - peroxisomal biogenesis factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000318.2 | 1191 | Missense Mutation | NP_000309.1 | |||
NM_001079867.1 | 1191 | Missense Mutation | NP_001073336.1 | |||
NM_001172086.1 | 1191 | Missense Mutation | NP_001165557.1 | |||
NM_001172087.1 | 1191 | Missense Mutation | NP_001165558.1 |