Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCATATGGACAGGCCATACCACGC[A/G]GAAGCCAGCCCTTCCCTGAAGCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615093 | ||||||||||||||||||||
Literature Links: |
LOC100288181 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100288181 - uncharacterized LOC100288181 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LY6K - lymphocyte antigen 6 complex, locus K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160354.1 | 684 | Silent Mutation | GCA,GCG | A,A 89 | NP_001153826.1 | |
NM_001160355.1 | 684 | Intron | NP_001153827.1 | |||
NM_017527.3 | 684 | Intron | NP_059997.3 | |||
XM_005250989.4 | 684 | Intron | XP_005251046.1 | |||
XM_005250990.4 | 684 | UTR 3 | XP_005251047.1 |