Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTAGACATCTGTTTCCAGAGCTTC[A/G]ACAATGTGTGCAGCAGAAGGACGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609353 MIM: 611210 | ||||||||||||||||||||
Literature Links: |
ESCO2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ESCO2 - establishment of sister chromatid cohesion N-acetyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017420.2 | 1384 | Intron | NP_001017420.1 | |||
XM_011544421.2 | 1384 | Intron | XP_011542723.1 | |||
XM_011544422.2 | 1384 | Intron | XP_011542724.1 |
PBK - PDZ binding kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278945.1 | 1384 | Silent Mutation | NP_001265874.1 | |||
NM_018492.3 | 1384 | Silent Mutation | NP_060962.2 | |||
XM_006716368.2 | 1384 | Silent Mutation | XP_006716431.1 |