Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGGTGTTTGTTGCTGGTTTCCT[C/G]CTTCTCGATCAACTCTAAGATGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607553 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPPK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPPK1 - epiplakin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031308.3 | 6736 | Missense Mutation | CAG,GAG | Q,E 2165 | NP_112598.3 | |
XM_017013889.1 | 6736 | Missense Mutation | CAG,GAG | Q,E 2198 | XP_016869378.1 | |
XM_017013890.1 | 6736 | Missense Mutation | CAG,GAG | Q,E 2165 | XP_016869379.1 | |
XM_017013891.1 | 6736 | Missense Mutation | CAG,GAG | Q,E 1237 | XP_016869380.1 | |
XM_017013892.1 | 6736 | Missense Mutation | CAG,GAG | Q,E 2198 | XP_016869381.1 |