Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGAACTCCCTGGGGTGAAGAAGT[A/G]TGAAGTGGTTTATCCTATAAGACTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606188 | ||||||||||||||||||||
Literature Links: |
ADAM28 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAM28 - ADAM metallopeptidase domain 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304351.1 | 193 | Missense Mutation | TAT,TGT | Y,C 28 | NP_001291280.1 | |
NM_014265.5 | 193 | Missense Mutation | TAT,TGT | Y,C 28 | NP_055080.2 | |
NM_021777.4 | 193 | Missense Mutation | TAT,TGT | Y,C 28 | NP_068547.2 | |
XM_005273380.3 | 193 | Missense Mutation | TAT,TGT | Y,C 28 | XP_005273437.1 | |
XM_005273382.1 | 193 | Missense Mutation | TAT,TGT | Y,C 28 | XP_005273439.1 | |
XM_006716273.3 | 193 | Missense Mutation | TAT,TGT | Y,C 28 | XP_006716336.1 | |
XM_006716274.1 | 193 | Missense Mutation | TAT,TGT | Y,C 28 | XP_006716337.1 | |
XM_011544367.2 | 193 | Missense Mutation | TAT,TGT | Y,C 28 | XP_011542669.1 | |
XM_011544368.2 | 193 | Missense Mutation | TAT,TGT | Y,C 28 | XP_011542670.1 | |
XM_011544369.2 | 193 | Missense Mutation | TAT,TGT | Y,C 28 | XP_011542671.1 | |
XM_011544370.2 | 193 | UTR 5 | XP_011542672.1 | |||
XM_011544371.2 | 193 | Intron | XP_011542673.1 | |||
XM_017012974.1 | 193 | Missense Mutation | TAT,TGT | Y,C 28 | XP_016868463.1 | |
XM_017012975.1 | 193 | Missense Mutation | TAT,TGT | Y,C 28 | XP_016868464.1 | |
XM_017012976.1 | 193 | UTR 5 | XP_016868465.1 |
LOC101929294 - uncharacterized LOC101929294 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |