Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAATGGAGGCGAGAAGGCCTCAG[A/T]GAAACTCGCCCCAGAAGAAGTTCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C8orf33 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C8orf33 - chromosome 8 open reading frame 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023080.2 | 329 | Missense Mutation | GAG,GTG | E,V 92 | NP_075568.1 |
LOC107983994 - endogenous retrovirus group K member 9 Pol protein-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |