Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTACCTGAATAAGTGATGTTTTA[C/T]CATGGTCCTTGCGATGCCGATAAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611941 | ||||||||||||||||||||
Literature Links: |
ATAD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATAD2 - ATPase family, AAA domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014109.3 | 2640 | Missense Mutation | NP_054828.2 | |||
XM_011516994.2 | 2640 | Missense Mutation | XP_011515296.1 | |||
XM_011516995.2 | 2640 | Intron | XP_011515297.1 | |||
XM_011516996.2 | 2640 | Missense Mutation | XP_011515298.1 |