Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCAAAAGGCCATTCCACCATTC[C/T]GTCAAGCCAGCTGCTTTAGAACGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FER1L6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FER1L6 - fer-1 like family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039112.2 | 47 | UTR 5 | NP_001034201.2 | |||
XM_006716618.3 | 47 | Intron | XP_006716681.1 | |||
XM_011517231.2 | 47 | Intron | XP_011515533.1 | |||
XM_011517232.2 | 47 | Intron | XP_011515534.1 | |||
XM_011517233.2 | 47 | Intron | XP_011515535.1 | |||
XM_011517235.2 | 47 | Intron | XP_011515537.1 | |||
XM_011517236.2 | 47 | Intron | XP_011515538.1 |