Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCCCGGTGGATGAAGTCGGCAGC[C/T]GTCTGGAAGTGGATGCTCATGTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUSP26 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DUSP26 - dual specificity phosphatase 26 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305115.1 | 678 | Silent Mutation | ACA,ACG | T,T 132 | NP_001292044.1 | |
NM_001305116.1 | 678 | Silent Mutation | ACA,ACG | T,T 132 | NP_001292045.1 | |
NM_024025.2 | 678 | Silent Mutation | ACA,ACG | T,T 132 | NP_076930.1 |