Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTCAGTCACCCCACTGGTTTCAG[C/G]GGCACATTACTCACTACCACCTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 193002 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC18A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC18A1 - solute carrier family 18 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135691.2 | 1977 | UTR 3 | NP_001129163.1 | |||
NM_001142324.1 | 1977 | UTR 3 | NP_001135796.1 | |||
NM_001142325.1 | 1977 | UTR 3 | NP_001135797.1 | |||
NM_003053.3 | 1977 | UTR 3 | NP_003044.1 | |||
XM_011544625.1 | 1977 | UTR 3 | XP_011542927.1 | |||
XM_011544626.1 | 1977 | UTR 3 | XP_011542928.1 | |||
XM_011544627.1 | 1977 | UTR 3 | XP_011542929.1 |