Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCAGGAGTCCCCTTCTGTCCCGT[C/T]CCCTGTCAGTCTCTCATCACCGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 165280 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLI4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLI4 - GLI family zinc finger 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138465.3 | 129 | Missense Mutation | TCC,TTC | S,F 15 | NP_612474.1 |
ZFP41 - ZFP41 zinc finger protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |