Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACCTGTCCCTCCAAAAGGACTG[G/T]CTTACAAATTTTCGTCCAAAAATGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605417 MIM: 174760 | ||||||||||||||||||||
Literature Links: |
DKK4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DKK4 - dickkopf WNT signaling pathway inhibitor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014420.2 | 911 | Missense Mutation | ACA,CCA | T,P 168 | NP_055235.1 | |
XM_011544488.2 | 911 | Missense Mutation | ACA,CCA | T,P 168 | XP_011542790.1 | |
XM_017013316.1 | 911 | Missense Mutation | ACA,CCA | T,P 168 | XP_016868805.1 |
POLB - DNA polymerase beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |