Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCGCCTCCTCCGGGCTATCTGTCA[C/G]GTGGCCTTTACTCTGCCGTCGGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TDRP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TDRP - testis development related protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256113.1 | 1099 | Intron | NP_001243042.1 | |||
NM_175075.4 | 1099 | Missense Mutation | CTG,GTG | L,V 177 | NP_778250.2 | |
XM_006716232.3 | 1099 | Missense Mutation | CTG,GTG | L,V 177 | XP_006716295.1 | |
XM_017013113.1 | 1099 | Missense Mutation | CTG,GTG | L,V 187 | XP_016868602.1 |