Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGCGGTTTTCACAGCATTAGGTG[A/C]ATGGGAAATGCCATCCAAATTCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601509 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GGH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GGH - gamma-glutamyl hydrolase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003878.2 | 1064 | Missense Mutation | GCA,TCA | A,S 261 | NP_003869.1 | |
XM_011517623.2 | 1064 | Intron | XP_011515925.1 |