Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTAGTTGGTAACCTTGGTGTACAC[A/G]CCCGGGACATCCTTCTGTCCACAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610469 MIM: 173370 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP3M2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP3M2 - adaptor related protein complex 3 mu 2 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLAT - plasminogen activator, tissue type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000930.4 | 1751 | Silent Mutation | GGC,GGT | G,G 544 | NP_000921.1 | |
NM_001319189.1 | 1751 | Silent Mutation | GGC,GGT | G,G 455 | NP_001306118.1 | |
NM_033011.3 | 1751 | Silent Mutation | GGC,GGT | G,G 498 | NP_127509.1 |