Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGTCATGGATTCATAAATACATA[A/T]GTATTTTGTACACAATGTGCTTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607352 MIM: 603612 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RHOBTB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RHOBTB2 - Rho related BTB domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160036.1 | 4016 | UTR 3 | NP_001153508.1 | |||
NM_001160037.1 | 4016 | UTR 3 | NP_001153509.1 | |||
NM_015178.2 | 4016 | UTR 3 | NP_055993.2 | |||
XM_017013251.1 | 4016 | UTR 3 | XP_016868740.1 |
TNFRSF10B - TNF receptor superfamily member 10b | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003842.4 | 4016 | UTR 3 | NP_003833.4 | |||
NM_147187.2 | 4016 | UTR 3 | NP_671716.2 | |||
XM_017013944.1 | 4016 | UTR 3 | XP_016869433.1 |